Invitrogen
Manufacturer Code:PA577522
Catalog # PIPA577522
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)KVPSTDITLRPTRK corresponding to amino acid residues 648-661 of rat Neuroligin 1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Neuroligin I; Neuroligin-1; Nlgn1
Gene Aliases: 6330415N05Rik; BB179718; Kiaa1070; mKIAA1070; NL1; Nlg1; Nlgn1
UniProt ID: (Mouse) Q99K10, (Rat) Q62765
Entrez Gene ID: (Mouse) 192167, (Rat) 116647
Molecular Function:
esterase
hydrolase
lipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.