Novus Biologicals
Manufacturer Code:NBP289932JF549
Catalog # NB110218
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 79 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human Neuroligin 1/NLGN1 (NP_055747.1 Met1-Ser677). |
Conjugate | Janelia Fluor 549 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 3.1.1 EC 3.1.1.1 KIAA1070neuroligin-1 MGC45115 neuroligin 1 NL1; Neuroligin-1
Gene Aliases: KIAA1070; NL1; NLGN1
UniProt ID: (Human) Q8N2Q7
Entrez Gene ID: (Human) 22871
Molecular Function: esterase hydrolase lipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.