Novus Biologicals
Manufacturer Code:NB1001881
Catalog # NB1001881
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-TAKQDDPKQQ corresponding to internal region according to NP_055747.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: EC 3.1.1 EC 3.1.1.1 KIAA1070neuroligin-1 MGC45115 neuroligin 1 NL1; Neuroligin-1
Gene Aliases: KIAA1070; NL1; NLGN1
UniProt ID: (Human) Q8N2Q7
Entrez Gene ID: (Human) 22871
Molecular Function:
esterase
hydrolase
lipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.