Invitrogen
Manufacturer Code:MA527635
Catalog # PIMA527635
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S97A-31 |
Immunogen | Fusion protein amino acids 718-843 (cytoplasmic C-terminus) of rat Neuroligin-1. Mouse: 99&percnt identity (125/126 amino acids identical). Human: 99&percnt identity (125/126 amino acids identical) >40&percnt identity with Neuroligin-2 and -3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Neuroligin I; Neuroligin-1; NLG 1 KIAA1070 MGC45115 Neuroligin-1 NL1 NLG1 Nlgn1
Gene Aliases: 6330415N05Rik; BB179718; KIAA1070; mKIAA1070; NL1; Nlg1; NLGN1
UniProt ID: (Human) Q8N2Q7, (Mouse) Q99K10, (Rat) Q62765
Entrez Gene ID: (Human) 22871, (Mouse) 192167, (Rat) 116647
Molecular Function: esterase hydrolase lipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.