Invitrogen
Manufacturer Code:PA136126
Catalog # 01-672-284
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Neurokinin A/Substance K conjugated to BSA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-terminal-flanking peptide; Neurokinin A; Neuromedin L; Neuropeptide gamma; Neuropeptide K; NKA; NPK; PPT; PPTA3 Ppt5fl RATPPTA3 TAC; Protachykinin-1; Substance K; Substance P; Tachykinin (substance P, neurokinin A, neuropeptide K, neuropeptide gamma); tachykinin 1
Gene Aliases: Nka; Nkna; Ppt5fl; PPTA3; RATPPTA3; TAC; Tac1; Tac2
UniProt ID: (Rat) P06767
Entrez Gene ID: (Rat) 24806
Molecular Function: neuropeptide peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.