Invitrogen
Manufacturer Code:PA591933
Catalog # PIPA591933
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human NEUROG1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bHLHa6; Class A basic helix-loop-helix protein 6; Helix-loop-helix protein mATH-4C; mATH4C; NeuroD3; Neurogenic basic-helix-loop-helix protein; neurogenic differentiation 3; Neurogenic differentiation factor 3; Neurogenin-1; NGN-1
Gene Aliases: AKA; Ath4c; BHLHA6; Math4C; NEUROD3; NEUROG1; NGN; NGN1
UniProt ID: (Human) Q92886, (Mouse) P70660
Entrez Gene ID: (Human) 4762, (Mouse) 18014
Molecular Function:
basic helix-loop-helix transcription factor
nuclease
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.