Invitrogen
Manufacturer Code:PA572637
Catalog # PIPA572637
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 112-138 amino acids from the Central region of DANRE neurog1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: basic helix-loop-helix transcription factor; NeuroD3; neurogenic differentiation 3; Neurogenic differentiation factor 3; Neurogenin-1; Neurogenin-1 NGN-1 Neurogenic differentiation factor 3 NeuroD3 Neurogenin-related protein 1 neurog1 neurod3 ngn1 ngr1; Neurogenin-related protein 1; NGN-1; transcription factor
Gene Aliases: cb260; chunp6899; neurod3; neurog1; NGN-1; ngn1; ngr1; Zath4c; zNgn1
UniProt ID: (Zebrafish) O42606
Entrez Gene ID: (Zebrafish) 30239
Molecular Function: basic helix-loop-helix transcription factor nuclease nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.