Invitrogen
Manufacturer Code:PA540478
Catalog # PIPA540478
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human NEUROG1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bHLHa6; Class A basic helix-loop-helix protein 6; Helix-loop-helix protein mATH-4C; mATH4C; NeuroD3; Neurogenic basic-helix-loop-helix protein; neurogenic differentiation 3; Neurogenic differentiation factor 3; Neurogenin 1 AKA Math4C NEUROD3 ngn1 bHLHa6; Neurogenin-1; NGN-1
Gene Aliases: AKA; ATH4C; BHLHA6; Math4C; NEUROD3; NEUROG1; NGN; NGN1
UniProt ID: (Human) Q92886, (Mouse) P70660, (Rat) P70595
Entrez Gene ID: (Human) 4762, (Pig) 100511625, (Rabbit) 100342109, (Dog) 481516, (Mouse) 18014, (Rat) 29410, (Guinea pig) 100715899
Molecular Function: basic helix-loop-helix transcription factor nuclease nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.