Invitrogen
Manufacturer Code:PA5117411
Catalog # PIPA5117411
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the C-terminus region of mouse Neurogenin 1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AKA Ath4c basic helix-loop-helix transcription factor BHLHA6 cb260 chunp6899 Class A basic helix-loop-helix protein 6 Helix-loop-helix protein mATH-4C Math4C neurod3 Neurog1 neurogenic basic-helix-loop-helix protein neurogenic differentiation 3 Neurogenic differentiation factor 3 neurogenin neurogenin 1 neurogenin related protein-1 neurogenin-1 Neurogenin-related protein 1 NGN ngn1 NGN-1 ngr1 transcription factor Zath4c zNgn1; Helix-loop-helix protein mATH-4C; mATH4C; NeuroD3; Neurogenic basic-helix-loop-helix protein; neurogenic differentiation 3; Neurogenic differentiation factor 3; Neurogenin-1; NGN-1
Gene Aliases: AKA; Ath4c; bHLHa6; Math4C; Neurod3; Neurog1; Ngn; Ngn1
UniProt ID: (Mouse) P70660, (Rat) P70595
Entrez Gene ID: (Mouse) 18014, (Rat) 29410
Molecular Function:
basic helix-loop-helix transcription factor
nuclease
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.