Novus Biologicals
Manufacturer Code:NB300154G
Catalog # NB300154G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | McNFn27b |
Immunogen | A highly conserved N-terminal peptide corresponding to amino acids 27-41 of human Neurofibromin 1. [UniProt# P21359] |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DKFZp686J1293 FLJ21220 Neurofibromatosis-related protein NF-1 neurofibromin neurofibromin 1 NFNS VRNF WSS; Neurofibromatosis-related protein NF-1; Neurofibromin; Neurofibromin truncated
Gene Aliases: NF1; NFNS; VRNF; WSS
UniProt ID: (Human) P21359
Entrez Gene ID: (Human) 4763
Molecular Function: G-protein modulator enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.