R&D Systems
Manufacturer Code:FAB8208S100UG
Catalog # FAB8208S100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1018843 |
Immunogen | Human embryonic kidney cell HEK293-derived human Neurofascin Ile25-Trp1039 Accession &num NP_055905 |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8°C as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DKFZp686P2250 KIAA0756FLJ46866 neurofascin neurofascin homolog neurofascin homolog (chicken) NF NFASC NRCAML; Neurofascin; neurofascin homolog
Gene Aliases: KIAA0756; NF; NFASC; NRCAML
UniProt ID: (Human) O94856
Entrez Gene ID: (Human) 23114
Molecular Function: G-protein coupled receptor cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.