R&D Systems
Manufacturer Code:AF3235
Catalog # AF3235
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant rat Neurofascin Ile25-Ala1031 Accession # NP_446361 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Chicken Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp686P2250 KIAA0756FLJ46866 neurofascin neurofascin homolog neurofascin homolog (chicken) NF NFASC NRCAML; Neurofascin; neurofascin homolog
Gene Aliases: KIAA0756; NF; NFASC; NRCAML
UniProt ID: (Human) O94856
Entrez Gene ID: (Human) 23114
Molecular Function:
G-protein coupled receptor
cell adhesion molecule
cytokine receptor
defense/immunity protein
hydrolase
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.