Proteintech
Manufacturer Code:263511AP150UL
Catalog # 50-173-5312
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Neurofascin Fusion Protein Ag24165 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA0756 Neurofascin neurofascin homolog (chicken) NF NFASC NRCAML; Neurofascin; neurofascin homolog
Gene Aliases: AA387016; D430023G06Rik; KIAA0756; mKIAA0756; NF; NFASC; NRCAML
UniProt ID: (Human) O94856, (Rat) P97685, (Mouse) Q810U3
Entrez Gene ID: (Human) 23114, (Rat) 116690, (Mouse) 269116
Molecular Function:
G-protein coupled receptor
cell adhesion molecule
cytokine receptor
defense/immunity protein
hydrolase
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.