R&D Systems
Manufacturer Code:AF6508SP
Catalog # AF6508SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human Neurocan Glu23-Cys1321 Accession # O14594 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: Chondroitin sulfate proteoglycan 3; chondroitin sulfate proteoglycan 3 (neurocan); Chondroitin sulfate proteoglycan 3 chondroitin sulfate proteoglycan 3 (neurocan) CSPG3 CSPG3neurocan core protein FLJ44681 NCAN NEUR neurocan neurocan proteoglycan; Neurocan core protein; neurocan proteoglycan
Gene Aliases: CSPG3; NCAN; NEUR
UniProt ID: (Human) O14594
Entrez Gene ID: (Human) 1463
Molecular Function: extracellular matrix glycoprotein extracellular matrix protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.