Invitrogen
Manufacturer Code:PA537188
Catalog # PIPA537188
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues in human Neurensin-2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Neurensin 2 Neurensin2 NRSN2 C20orf98; Neurensin-2; nrsn2
Gene Aliases: C20orf98; dJ1103G7.6; Gm123; Neurensin-2; NRSN2
UniProt ID: (Human) Q9GZP1, (Mouse) Q5HZK2
Entrez Gene ID: (Human) 80023, (Mouse) 228777, (Rat) 689978
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.