Novus Biologicals
Manufacturer Code:NBP159338
Catalog # NBP159338
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to NRG3(neuregulin 3) The peptide sequence was selected from the middle region of NRG3. Peptide sequence TSINMQLPSRETNPYFNSLEQKDLVGYSSTRASSVPIIPSVGLEETCLQM. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HRG3 neuregulin 3 neuregulin 3 variant 1 neuregulin 3 variant 10 neuregulin 3 variant 11 neuregulin 3 variant 12 neuregulin 3 variant 13 neuregulin 3 variant 14 neuregulin 3 variant 3 neuregulin 3 variant 4 neuregulin 3 variant 5 neuregulin 3 variant 6 neuregulin 3 variant 7 neuregulin 3 variant 8 neuregulin 3 variant 9 neuregulin-3-like polypeptide pro-neuregulin-3 membrane-bound isoform pro-NRG3; neuregulin 3 variant 1; neuregulin 3 variant 10; neuregulin 3 variant 11; neuregulin 3 variant 12; neuregulin 3 variant 13; neuregulin 3 variant 14; neuregulin 3 variant 3; neuregulin 3 variant 4; neuregulin 3 variant 5; neuregulin 3 variant 6; neuregulin 3 variant 7; neuregulin 3 variant 8; neuregulin 3 variant 9; Neuregulin-3; neuregulin-3-like polypeptide; NRG-3; Pro-neuregulin-3, membrane-bound isoform; Pro-NRG3
Gene Aliases: HRG3; NRG3; pro-NRG3
UniProt ID: (Human) P56975
Entrez Gene ID: (Human) 10718
Molecular Function: enzyme modulator growth factor kinase activator kinase modulator membrane-bound signaling molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.