R&D Systems
Manufacturer Code:AF296SP
Catalog # AF296SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human NRG1-alpha/HRG1-alpha Ser177-Lys241 Accession # Q7RTV8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Acetylcholine receptor-inducing activity; ARIA; ARIA GGF2 Heregulin-1 HGL HRG HRG1 HRG1-alpha HRG1-beta 1 MST131 MSTP131 neuregulin 1 Neuregulin1 Neuregulin1 alpha NRG1 pro-neuregulin-1 membrane-bound isoform pro-NRG1 sensory and motor neuron derived factor SMDF; Breast cancer cell differentiation factor p45; Glial growth factor; glial growth factor 2; Heregulin; heregulin, alpha (45kD, ERBB2 p185-activator); HRG; Neu differentiation factor; Neuregulin-1; Pro-neuregulin-1, membrane-bound isoform; Pro-NRG1; sensory and motor neuron derived factor; Sensory and motor neuron-derived factor
Gene Aliases: ARIA; GGF; GGF2; HGL; HRG; HRG1; HRGA; MST131; MSTP131; NDF; NRG1; NRG1-IT2; SMDF
UniProt ID: (Human) A5YAK5
Entrez Gene ID: (Human) 3084
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.