Novus Biologicals
Manufacturer Code:NBP287901
Catalog # NB035523
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of Human Neurabin 1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA1222NRBI neurabin-1 Neurabin-IFLJ20068 Neural tissue-specific F-actin-binding protein I NRB1 Protein phosphatase 1 regulatory subunit 9A protein phosphatase 1 regulatory (inhibitor) subunit 9A; Neurabin-1; Neurabin-I; Neural tissue-specific F-actin-binding protein I; Protein phosphatase 1 regulatory subunit 9A; protein phosphatase 1, regulatory (inhibitor) subunit 9A; protein phosphatase 1, regulatory subunit 9A
Gene Aliases: KIAA1222; Neurabin-I; NRB1; NRBI; PPP1R9A
UniProt ID: (Human) Q9ULJ8
Entrez Gene ID: (Human) 55607
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.