Proteintech
Manufacturer Code:601311IG150UL
Catalog # 50-173-6257
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4G9E12 |
Immunogen | Neudesin/NENF Fusion Protein Ag8387 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: cell growth-inhibiting protein 47; Cell immortalization-related protein 2; CIR2 NENF NEUDESIN SCIRP10 SPUF SPUF protein; Neudesin; Neuron-derived neurotrophic factor; Protein GIG47; SCIRP10-related protein; Secreted protein of unknown function; Spinal cord injury related protein 10; SPUF protein
Gene Aliases: 1110060M21Rik; CIR2; NENF; SCIRP10; SPUF
UniProt ID: (Human) Q9UMX5, (Mouse) Q9CQ45
Entrez Gene ID: (Human) 29937, (Mouse) 66208
Molecular Function: receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.