Invitrogen
Manufacturer Code:PA578639
Catalog # PIPA578639
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of Mouse NeuN. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Fox-1 homolog C; Fox-3; Fox3 Hrnbp3 Neuna60 RNA binding protein fox-1 homolog (C. elegans) 3 Rbfox3; hexaribonucleotide binding protein 3; Hexaribonucleotide-binding protein 3; NeuN antigen; neuronal nuclear antigen A60; neuronal nuclei; Neuronal nuclei antigen; RNA binding protein fox-1 homolog 3; RNA binding protein, fox-1 homolog 3
Gene Aliases: D11Bwg0517e; Fox-3; Hrnbp3; NeuN; Neuna60; Rbfox3; RGD1560070
UniProt ID: (Mouse) Q8BIF2
Entrez Gene ID: (Rat) 287847, (Mouse) 52897
Molecular Function: RNA binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.