Invitrogen
Manufacturer Code:PA556560
Catalog # PIPA556560
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human NeuN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Fox-1 homolog C; hexaribonucleotide binding protein 3; NeuN antigen; neuronal nuclei; Neuronal nuclei antigen; RNA binding protein fox-1 homolog 3; RNA binding protein fox-1 homolog 3 Fox-1 homolog C Hexaribonucleotide-binding protein 3 Fox-3 Neuronal nuclei NeuN Rbfox3 D11Bwg0517e Hrnbp3; RNA binding protein, fox-1 homolog (C. elegans) 3
Gene Aliases: FOX-3; FOX3; HRNBP3; NEUN; RBFOX3
UniProt ID: (Human) A6NFN3
Entrez Gene ID: (Human) 146713
Molecular Function: RNA binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.