Invitrogen
Manufacturer Code:PA5115641
Catalog # PIPA5115641
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NeuN corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D11Bwg0517e Fox-1 homolog C FOX3 Fox-3 hexaribonucleotide binding protein 3 Hexaribonucleotide-binding protein 3 Hrnbp3 Neun NeuN antigen Neuna60 neuronal nuclear antigen A60 neuronal nuclei Neuronal nuclei antigen Rbfox3 RFOX3 RGD1560070 RNA binding fox-1 homolog 3 RNA binding protein fox-1 homolog 3 RNA binding protein fox-1 homolog (C. elegans) 3 RNA binding protein fox-1 homolog 3; Fox-1 homolog C; Fox-3; hexaribonucleotide binding protein 3; Hexaribonucleotide-binding protein 3; NeuN antigen; neuronal nuclear antigen A60; neuronal nuclei; Neuronal nuclei antigen; RNA binding protein fox-1 homolog 3; RNA binding protein, fox-1 homolog (C. elegans) 3; RNA binding protein, fox-1 homolog 3
Gene Aliases: D11Bwg0517e; FOX-3; FOX3; HRNBP3; NEUN; Neuna60; RBFOX3; RGD1560070
UniProt ID: (Human) A6NFN3, (Mouse) Q8BIF2
Entrez Gene ID: (Human) 146713, (Rat) 287847, (Mouse) 52897
Molecular Function: RNA binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.