R&D Systems
Manufacturer Code:MAB1254SP
Catalog # MAB1254SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 191633 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Netrin-4 Val26-Lys628 Accession # Q9HB63 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Beta-netrin; beta-netrin hepar-derived netrin-like protein netrin 4 Netrin4 NTN4 PRO3091; Hepar-derived netrin-like protein; Netrin-4
Gene Aliases: NTN4; PRO3091
UniProt ID: (Human) Q9HB63
Entrez Gene ID: (Human) 59277
Molecular Function:
extracellular matrix linker protein
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.