Novus Biologicals
Manufacturer Code:NBP259944JF646
Catalog # NBP59944646
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 6C11 |
Immunogen | GST fused to residues 6653-6835 of human Nesp2 |
Conjugate | Janelia Fluor 646 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: DKFZP434H2235 DKFZp686E01115 DKFZp686H1931 EDMD5 FLJ46790 KIAA1011FLJ11014 nesprin 2 nesprin-2 NUAFLJ43727 NUANCEFLJ45710 Nuclear envelope spectrin repeat protein 2 Nucleus and actin connecting element protein polytrophin Protein NUANCE spectrin repeat containing nuclear envelope 2 Synaptic nuclear envelope protein 2 synaptic nuclei expressed gene 2 SYNE-2 TROPH; KASH domain-containing protein 2; KASH2; nesprin 2; Nesprin-2; Nuclear envelope spectrin repeat protein 2; Nucleus and actin connecting element protein; polytrophin; Protein NUANCE; spectrin repeat containing, nuclear envelope 2; Synaptic nuclear envelope protein 2; synaptic nuclei expressed gene 2; Syne-2
Gene Aliases: EDMD5; KIAA1011; Nesp2; Nesprin-2; NUA; NUANCE; SYNE-2; SYNE2; TROPH
UniProt ID: (Human) Q8WXH0
Entrez Gene ID: (Human) 23224
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.