Novus Biologicals
Manufacturer Code:NBP159767
Catalog # NBP159767
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to Nesprin-4 The peptide sequence was selected from the N terminal of Nesprin-4. Peptide sequence GEESTSPEQAQTLGQDSLGPPEHFQGGPRGNEPAAHPPRWSTPSSYEDPA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: chromosome 19 open reading frame 46 FLJ36445 nesprin-4; deafness, autosomal recessive 76; KASH domain-containing protein 4; KASH4; Nesprin-4; Nuclear envelope spectrin repeat protein 4; spectrin repeat containing, nuclear envelope family member 4
Gene Aliases: C19orf46; DFNB76; Nesp4; SYNE4
UniProt ID: (Human) Q8N205
Entrez Gene ID: (Human) 163183
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.