Invitrogen
Manufacturer Code:MA534670
Catalog # PIMA534670
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JB35-26 |
Immunogen | Recombinant protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles store in dark |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 8B A330049M09Rik ARCA1 BE692247 C130039F11Rik C6orf98 CPG2 dJ45H2.2 DKFZp781J13156 EDMD4 Enaptin FLJ30878 FLJ41140 KASH domain-containing protein 1 KASH1 KIAA0796 KIAA1262 KIAA1756 mKIAA1756 MYNE1 Myne-1 Myocyte nuclear envelope protein 1 Nesp1 Nesprin-1 Nuclear envelope spectrin repeat protein 1 RP1-130E4.2 SCAR8 Synaptic nuclear envelope protein 1 SYNE 1 Syne1 Syne-1 syne-1 fragment; CPG2 full length; Enaptin; KASH domain-containing protein 1; KASH1; Myne-1; Myocyte nuclear envelope protein 1; nesprin 1; Nesprin-1; Nuclear envelope spectrin repeat protein 1; spectrin repeat containing, nuclear envelope 1; Synaptic nuclear envelope protein 1; synaptic nuclei expressed gene 1; Syne-1
Gene Aliases: 8B; ARCA1; C6orf98; CPG2; dJ45H2.2; EDMD4; KIAA0796; KIAA1262; KIAA1756; MYNE1; Nesp1; SCAR8; SYNE1
UniProt ID: (Human) Q8NF91
Entrez Gene ID: (Human) 23345
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.