R&D Systems
Manufacturer Code:MAB8025SP
Catalog # MAB8025SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 859326 |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human Nephronectin Glu20-Arg565 Accession # Q6UXI9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EGFL6L Nctn nephronectin NPNT POEM; Nephronectin; Preosteoblast EGF-like repeat protein with MAM domain; Protein EGFL6-like
Gene Aliases: EGFL6L; NPNT; POEM; UNQ295/PRO334
UniProt ID: (Human) Q6UXI9
Entrez Gene ID: (Human) 255743
Molecular Function:
extracellular matrix protein
extracellular matrix structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.