Invitrogen
Manufacturer Code:PA572021
Catalog # PIPA572021
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 657-685 amino acids from the Central region of human NEO1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Immunoglobulin superfamily DCC subclass member 2; NEO NEO1 NGN HsT17534 IGDCC2DKFZp547B146 Immunoglobulin superfamily DCC subclass member 2 neogenin neogenin (chicken) homolog 1 neogenin 1 neogenin homolog 1 NGNDKFZp547A066; Neogenin; neogenin homolog 1
Gene Aliases: 2610028H22Rik; AI327052; D930014N22Rik; IGDCC2; NEO1; NGN; NTN1R2
UniProt ID: (Human) Q92859
Entrez Gene ID: (Human) 4756, (Mouse) 18007
Molecular Function: G-protein coupled receptor cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.