R&D Systems
Manufacturer Code:AF1079SP
Catalog # AF1079SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse Neogenin Ala42-Ile1033 (Asp442-Leu461 del) Accession # NP_032710 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: HsT17534 IGDCC2DKFZp547B146 Immunoglobulin superfamily DCC subclass member 2 NEO NEO1 neogenin neogenin (chicken) homolog 1 neogenin 1 neogenin homolog 1 NGN NGNDKFZp547A066; Immunoglobulin superfamily DCC subclass member 2; Neogenin; neogenin homolog 1
Gene Aliases: IGDCC2; NEO1; NGN; NTN1R2
UniProt ID: (Human) Q92859
Entrez Gene ID: (Human) 4756
Molecular Function: G-protein coupled receptor cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.