Invitrogen
Manufacturer Code:PA595385
Catalog # PIPA595385
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the N-terminus of human Nectin-4/PVRL4 different from the related mouse sequence by seven amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ig superfamily receptor LNIR; nectin 4; Nectin cell adhesion molecule 4; Nectin-4; Nectin-4 Ig superfamily receptor LNIR Nectin cell adhesion molecule 4 Poliovirus receptor-related protein 4 Processed poliovirus receptor-related protein 4 NECTIN4 LNIR PRR4 PVRL4; poliovirus receptor-related 4; Poliovirus receptor-related protein 4; Processed poliovirus receptor-related protein 4
Gene Aliases: EDSS1; LNIR; nectin-4; NECTIN4; PRR4; PVRL4
UniProt ID: (Human) Q96NY8
Entrez Gene ID: (Human) 81607
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.