Invitrogen
Manufacturer Code:PA5110246
Catalog # PIPA5110246
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 371-510 of human PVRL4 (NP_112178.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1200017F15Rik EDSS1 Ig superfamily receptor LNIR Lnir Nectin cell adhesion molecule 4 Nectin4 Nectin-4 Poliovirus receptor-related protein 4 Processed poliovirus receptor-related protein 4 Prr4 PVRL4 RGD1559826; Ig superfamily receptor LNIR; nectin 4; Nectin cell adhesion molecule 4; Nectin-4; poliovirus receptor-related 4; Poliovirus receptor-related protein 4; Processed poliovirus receptor-related protein 4
Gene Aliases: 1200017F15Rik; EDSS1; LNIR; nectin-4; NECTIN4; PRR4; PVRL4; RGD1559826
UniProt ID: (Human) Q96NY8, (Mouse) Q8R007
Entrez Gene ID: (Human) 81607, (Mouse) 71740, (Rat) 498281
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.