Invitrogen
Manufacturer Code:MA533378
Catalog # PIMA533378
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Functional Assay (FN) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 103-A1 |
Immunogen | Recombit mouse nectin-3 extracellular domain (50-400 aa) fused to IgG Fc |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 2610301B19Rik 3000002N23Rik 4921513D19Rik AA407785 AU016832 AW538082; CD113; Nectin cell adhesion molecule 3; Nectin-3; Poliovirus receptor-related protein 3
Gene Aliases: 2610301B19Rik; 3000002N23Rik; 4921513D19Rik; AA407785; AU016832; AW538082; CD113; Nectin3; Pvrl3
UniProt ID: (Mouse) Q9JLB9
Entrez Gene ID: (Mouse) 58998
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.