R&D Systems
Manufacturer Code:FAB3869P
Catalog # FAB3869P
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 829038 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse Nectin-2/CD112 Gln32-Gly351 (predicted) Accession # P32507 |
Conjugate | R-PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CD112; CD112 CD112 antigen Herpes virus entry mediator B Herpesvirus entry mediator B herpesvirus entry protein B HVEB HVEBpoliovirus receptor-like 2 MPH nectin 2 Nectin2 Nectin-2 poliovirus receptor-related 2 (herpesvirus entry mediator B) poliovirus receptor-related protein 2 PRR2 PRR2nectin-2 PVRL2 PVRR2poliovirus receptor related 2; Herpes virus entry mediator B; Herpesvirus entry mediator B; herpesvirus entry protein B; Nectin cell adhesion molecule 2; Nectin-2; poliovirus receptor-like 2; poliovirus receptor-related 2 (herpesvirus entry mediator B); Poliovirus receptor-related protein 2
Gene Aliases: CD112; HVEB; NECTIN2; PRR2; PVRL2; PVRR2
UniProt ID: (Human) Q92692
Entrez Gene ID: (Human) 5819
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.