R&D Systems
Manufacturer Code:AF2229SP
Catalog # AF2229SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Nectin-2/CD112 isoform a Gln32-Leu360 Accession # NP_002847 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CD112; CD112 CD112 antigen Herpes virus entry mediator B Herpesvirus entry mediator B herpesvirus entry protein B HVEB HVEBpoliovirus receptor-like 2 MPH nectin 2 Nectin2 Nectin-2 poliovirus receptor-related 2 (herpesvirus entry mediator B) poliovirus receptor-related protein 2 PRR2 PRR2nectin-2 PVRL2 PVRR2poliovirus receptor related 2; Herpes virus entry mediator B; Herpesvirus entry mediator B; herpesvirus entry protein B; Nectin cell adhesion molecule 2; Nectin-2; poliovirus receptor-like 2; poliovirus receptor-related 2 (herpesvirus entry mediator B); Poliovirus receptor-related protein 2
Gene Aliases: CD112; HVEB; NECTIN2; PRR2; PVRL2; PVRR2
UniProt ID: (Human) Q92692
Entrez Gene ID: (Human) 5819
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.