Invitrogen
Manufacturer Code:MA535822
Catalog # PIMA535822
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1665 |
Immunogen | A synthesized peptide derived from human Nectin 2/CD112 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: AI325026 AI987993 CD112 Herpes virus entry mediator B Herpesvirus entry mediator B HVEB mHveB Mph Murine herpes virus entry protein B MYH2B MyHC-2B MyHC-IIb Nectin cell adhesion molecule 2 Nectin2 Nectin-2 Poliovirus receptor homolog Poliovirus receptor-related protein 2 PRR2 Pvr Pvrl2 PVRR2 Pvs sCD155 soluble CD155 soluble PVR sPVR; CD112; Herpes virus entry mediator B; Herpesvirus entry mediator B; herpesvirus entry protein B; Nectin cell adhesion molecule 2; Nectin-2; poliovirus receptor-like 2; poliovirus receptor-related 2 (herpesvirus entry mediator B); Poliovirus receptor-related protein 2
Gene Aliases: CD112; HVEB; NECTIN2; PRR2; PVRL2; PVRR2
UniProt ID: (Human) Q92692
Entrez Gene ID: (Human) 5819
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.