Invitrogen
Manufacturer Code:MA531080
Catalog # PIMA531080
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 13 |
Immunogen | Recombit Rat CD112/Nectin-2/PVRL2 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: CD112 HVEB PRR2 PVRR2 herpesvirus entry protein B nectin-2 poliovirus receptor-like 2 poliovirus receptor-related protein 2; nectin-2; poliovirus receptor related 2; poliovirus receptor-related 2 (herpesvirus entry mediator B); poliovirus receptor-related protein 2
Entrez Gene ID: (Rat) 308417
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.