Invitrogen
Manufacturer Code:PA569507
Catalog # PIPA569507
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human PVRL2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD112; Herpes virus entry mediator B; Herpesvirus entry mediator B; herpesvirus entry protein B; hveB; mHveB; Murine herpes virus entry protein B; murine herpesvirus entry protein B; Nectin cell adhesion molecule 2; Nectin-2; NECTIN2 HVEB PRR2 PVRL2; Poliovirus receptor homolog; poliovirus receptor related 2; poliovirus receptor-like 2; poliovirus receptor-related 2 (herpesvirus entry mediator B); Poliovirus receptor-related protein 2; poliovirus sensitivity
Gene Aliases: AI325026; AI987993; CD112; HVEB; MPH; nectin-2; NECTIN2; PRR2; Pvr; PVRL2; PVRR2; Pvs
UniProt ID: (Human) Q92692, (Mouse) P32507
Entrez Gene ID: (Human) 5819, (Pig) 100038012, (Mouse) 19294
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.