Invitrogen
Manufacturer Code:PA5115635
Catalog # PIPA5115635
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Necdin corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI528698 HsT16328 NDN necdin necdin homolog necdin MAGE family member necdin melanoma antigen (MAGE) family member necdin-like protein Peg6 Prader-Willi syndrome chromosome region PWCR; Necdin; necdin homolog; necdin, melanoma antigen (MAGE) family member; necdin-like protein; Prader-Willi syndrome chromosome region
Gene Aliases: AI528698; HsT16328; NDN; Peg6; PWCR
UniProt ID: (Human) Q99608, (Mouse) P25233
Entrez Gene ID: (Human) 4692, (Mouse) 17984, (Rat) 308690
Molecular Function: DNA binding protein cell adhesion molecule nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.