Abnova
Manufacturer Code:MAB2460
Catalog # 89-111-012
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Chinese hamster |
Class | Monoclonal |
Type | Antibody |
Clone | N1Nterm15-32.8 |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to mouse Ndfip1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Hamster monoclonal antibody raised against synthetic peptide of Ndfip1.
For Research Use Only
Protein Aliases: 0610010M22Rik; N4wbp5; NEDD4 family-interacting protein 1; Nedd4 WW binding protein 5; NEDD4 WW domain-binding protein 5; Nedd4 WW-binding protein 5
Gene Aliases: 0610010M22Rik; N4wbp5; Ndfip1
UniProt ID: (Mouse) Q8R0W6
Entrez Gene ID: (Mouse) 65113
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.