R&D Systems
Manufacturer Code:MAB1573
Catalog # MAB1573
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 206919 |
Immunogen | <EM>E. coli</EM>-derived recombinant human Nbs1 Pro498-Arg754 Accession # O60934 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ATV AT-V1 AT-V2 Cell cycle regulatory protein p95 FLJ10155 MGC87362 NBN NBS NBS1P95 nibrin Nijmegen breakage syndrome 1 (nibrin) Nijmegen breakage syndrome protein 1 p95 p95 protein of the MRE11/RAD50 complex; Cell cycle regulatory protein p95; Nibrin; Nijmegen breakage syndrome 1 (nibrin); Nijmegen breakage syndrome protein 1; p95 protein of the MRE11/RAD50 complex
Gene Aliases: AT-V1; AT-V2; ATV; NBN; NBS; NBS1; P95
UniProt ID: (Human) O60934
Entrez Gene ID: (Human) 4683
Molecular Function: DNA binding protein damaged DNA-binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.