Novus Biologicals
Manufacturer Code:NBP147615
Catalog # NBP147615
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S134-12 |
Immunogen | Fusion protein amino acids 1724-1956 of rat Nav1.8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: hPN3 Nav1.8 PN3 sodium channel protein type 10 subunit alpha sodium channel protein type X subunit alpha sodium channel voltage-gated type X alpha subunit voltage-gated type X alpha polypeptide; Peripheral nerve sodium channel 3; PN3; Sodium channel protein type 10 subunit alpha; Sodium channel protein type X subunit alpha; sodium channel, voltage gated, type X alpha subunit; sodium channel, voltage-gated, type X, alpha polypeptide; sodium channel, voltage-gated, type X, alpha subunit; Voltage-gated sodium channel subunit alpha Nav1.8
Gene Aliases: FEPS2; hPN3; Nav1.8; PN3; SCN10A; SNS
UniProt ID: (Human) Q9Y5Y9
Entrez Gene ID: (Human) 6336
Molecular Function:
calcium channel
ion channel
sodium channel
transporter
voltage-gated calcium channel
voltage-gated ion channel
voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.