Novus Biologicals
Manufacturer Code:NBP212904G
Catalog # NBP212904G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S68-6 |
Immunogen | Fusion protein amino acids 1751-1946 (C-terminus) of human Nav1.7 |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ETHA FEB3B NE-NA NENAVoltage-gated sodium channel subunit alpha Nav1.7 Neuroendocrine sodium channel Peripheral sodium channel 1 PN1hNE-Na sodium channel protein type 9 subunit alpha Sodium channel protein type IX subunit alpha sodium channel voltage-gated type IX alpha subunit voltage-gated sodium channel alpha subunit Nav1.7 voltage-gated type IX alpha polypeptide; hNE-Na; Neuroendocrine sodium channel; Peripheral sodium channel 1; PN1; Sodium channel protein type 9 subunit alpha; Sodium channel protein type IX subunit alpha; sodium channel, voltage gated, type IX alpha subunit; sodium channel, voltage-gated, type IX, alpha polypeptide; sodium channel, voltage-gated, type IX, alpha subunit; voltage-gated sodium channel alpha subunit Nav1.7; Voltage-gated sodium channel subunit alpha Nav1.7
Gene Aliases: ETHA; FEB3B; GEFSP7; HSAN2D; Nav1.7; NE-NA; NENA; PN1; SCN9A; SFNP
UniProt ID: (Human) Q15858
Entrez Gene ID: (Human) 6335
Molecular Function: calcium channel ion channel sodium channel transporter voltage-gated calcium channel voltage-gated ion channel voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.