Novus Biologicals
Manufacturer Code:NBP202401
Catalog # NBP202401
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4C3 |
Immunogen | Full length human recombinant protein of human NPR3 (NP_000899) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ANP-C; ANP-C ANPR-C ANPRCFLJ14054 Atrial natriuretic peptide clearance receptor atrial natriuretic peptide receptor 3 Atrial natriuretic peptide receptor type C C5orf23 chromosome 5 open reading frame 23 GUCY2B natriuretic peptide receptor C/guanylate cyclase C (atrionatriuretic peptidereceptor C) NPRC NPR-C; Atrial natriuretic peptide clearance receptor; Atrial natriuretic peptide receptor 3; Atrial natriuretic peptide receptor type C; atrionatriuretic peptide receptor C; guanylate cyclase C; natriuretic peptide receptor C/guanylate cyclase C (atrionatriuretic peptide receptor C)
Gene Aliases: ANP-C; ANPR-C; ANPRC; C5orf23; GUCY2B; NPR-C; NPR3; NPRC
UniProt ID: (Human) P17342
Entrez Gene ID: (Human) 4883
Molecular Function:
adenylate cyclase
cyclase
guanylate cyclase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.