Proteintech
Manufacturer Code:602591IG150UL
Catalog # 50-173-6356
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2D12A2 |
Immunogen | Napsin A Fusion Protein Ag9721 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: asp 4; Asp 4 ASP4 Aspartyl protease 4 KAP Kdap NAP1 NAPA NAPSA Napsin 1 Napsin A napsin A aspartic peptidase Pronapsin A SNAPA TA01/TA02; ASP4; Aspartyl protease 4; KAP; KDAP-1; Kidney-derived aspartic protease-like protein; Napsin-1; Napsin-A; pronapsin A; TA01/TA02
Gene Aliases: KAP; Kdap; Nap; NAP1; NAPA; NAPSA; pronapsin; SNAPA
UniProt ID: (Human) O96009, (Mouse) O09043
Entrez Gene ID: (Human) 9476, (Rat) 60575, (Mouse) 16541
Molecular Function:
aspartic protease
hydrolase
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.