Invitrogen
Manufacturer Code:PA520537
Catalog # PIPA520537
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 16 amino acid peptide near the carboxy terminus of the human os3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Nanos homolog 3; NANOS1L NOS3 os homolog 3; NOS-3
Gene Aliases: NANOS1L; NANOS3; NOS3; ZC2HC12C
UniProt ID: (Human) P60323, (Mouse) P60324
Entrez Gene ID: (Human) 342977, (Mouse) 244551, (Rat) 288909
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.