Novus Biologicals
Manufacturer Code:NBP304492
Catalog # NB095967
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-192 of human NANOS3 (NP_001092092.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MGC120114 nanos homolog 3 nanos homolog 3 (Drosophila) NOS-3 NOS3NANOS1L; Nanos homolog 3; NOS-3
Gene Aliases: NANOS1L; NANOS3; NOS3; ZC2HC12C
UniProt ID: (Human) P60323
Entrez Gene ID: (Human) 342977
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.