Invitrogen
Manufacturer Code:PA520557
Catalog # PIPA520557
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 14 amino acid peptide from near the center of human os1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC_Rep1a; nanos 1; Nanos homolog 1; nanos homolog 1 (Drosophila); NOS-1; RP11-498J9.3 NOS1 EC&lowbarRep1a NOS-1 os homolog 1
Gene Aliases: EC_Rep1a; NANOS1; Nos; NOS-1; NOS1; SPGF12; ZC2HC12A
UniProt ID: (Human) Q8WY41, (Mouse) Q80WY3
Entrez Gene ID: (Human) 340719, (Rat) 365494, (Mouse) 332397
Molecular Function: nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.