R&D Systems
Manufacturer Code:IC2729G
Catalog # IC2729G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant mouse Nanog Trp154-Leu262 Accession # Q80Z64 |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ12581 FLJ40451 hNanog homeobox protein NANOG Homeobox transcription factor Nanog homeobox transcription factor Nanog-delta 48 Nanog homeobox; hNanog; Homeobox protein NANOG; Homeobox transcription factor Nanog; homeobox transcription factor Nanog-delta 48
Gene Aliases: NANOG
UniProt ID: (Human) Q9H9S0
Entrez Gene ID: (Human) 79923
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.