Abnova
Manufacturer Code:MAB18373
Catalog # 89-338-298
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S187-7 |
Immunogen | Recombinant protein corresponding to amino acids 1659-1738 at C-terminus of rat Nalcn. |
Conjugate | ATTO594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: brain voltage-gated cation channel; Four domain-type voltage-gated ion channel alpha-1 subunit; Rb21-channel; Sodium leak channel non-selective protein; Vgcnl1; voltage gated channel like 1; Voltage gated channel-like protein 1
Gene Aliases: Nalcn; Nca; Rb21; Vgcnl1
UniProt ID: (Rat) Q6Q760
Entrez Gene ID: (Rat) 266760
Molecular Function:
calcium channel
ion channel
sodium channel
transporter
voltage-gated calcium channel
voltage-gated ion channel
voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.