Invitrogen
Manufacturer Code:MA514827
Catalog # PIMA514827
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | S.902.7 |
Immunogen | Synthetic peptide corresponding to residues surrounding Ala217 of human Naked2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Dvl-binding protein NKD2; Dvl-binding protein NKD2 hNkd2 naked cuticle-2 naked-2 protein naked cuticle homolog 2; Naked-2; Protein naked cuticle homolog 2
Gene Aliases: Naked2; NKD2
UniProt ID: (Human) Q969F2
Entrez Gene ID: (Human) 85409
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.